3,5-Difluoro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

19476-25G 19476-5G
76909-714EA 453.01 USD
76909-714 76909-716
3,5-Difluoro-4-methoxybenzaldehyde ≥97%
3,5-Difluoro-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04115911
CAS Number: 654-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR