4-Benzyloxy-2-methoxyphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

19397-1G 19397-25G 19397-500MG 19397-5G
76909-676EA 350.1 USD
76909-676 76909-678 76909-680 76909-682
4-Benzyloxy-2-methoxyphenylboronic acid ≥97% research grade
4-Benzyloxy-2-methoxyphenylboronic acid
Formula: C₁₄H₁₅BO₄
MW: 258.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22375025
CAS Number: 211495-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR