4-Bromo-2,6-difluoro-1-(benzyloxy)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
19339-1G
19339-25G
19339-5G
76909-662EA
66.84
USD
76909-662
76909-664
76909-666
4-Bromo-2,6-difluoro-1-(benzyloxy)benzene ≥97%
4-Bromo-2,6-difluoro-1-(benzyloxy)benzene
Formula:
C₁₃H₉BrF₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD16295263 CAS Number: 99045-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...