3-Bromo-5-fluorophenylboronic acid N-butyldiethanolamine ester ≥97%

Supplier: AOBChem USA

Warning

19327-25G 19327-5G
76909-656EA 204.76 USD
76909-656 76909-658
3-Bromo-5-fluorophenylboronic acid N-butyldiethanolamine ester ≥97%
3-Bromo-5-fluorophenylboronic acid N-butyldiethanolamine ester
Formula: C₁₄H₂₀BBrFNO₂
MW: 344.028 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543650
CAS Number: 1190989-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR