3-Bromo-2-methylthiophene ≥97%

Supplier: AOBChem USA

Warning

19148-25G 19148-5G
76909-604EA 257.8 USD
76909-604 76909-606
3-Bromo-2-methylthiophene ≥97%
3-Bromo-2-methylthiophene

Precaución: for Research Use Only

Formula: C₅H₅BrS
Storage Temperature: Ambient
MDL Number: MFCD09033008
CAS Number: 30319-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR