3,5-Dibromo-2-thiophenecarboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

19145-1G 19145-25G 19145-5G
76909-594EA 287.51 USD
76909-594 76909-596 76909-600
3,5-Dibromo-2-thiophenecarboxaldehyde ≥95%
3,5-Dibromo-2-thiophenecarboxaldehyde
Formula: C₅H₂Br₂OS
MW: 269.942 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426904
CAS Number: 23688-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR