3-Bromo-6-ethoxy-2-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

19137-1G 19137-25G 19137-5G
76909-584EA 62.6 USD
76909-584 76909-586 76909-588
3-Bromo-6-ethoxy-2-methylpyridine ≥97%
3-Bromo-6-ethoxy-2-methylpyridine
Formula: C₈H₁₀BrNO
MW: 216.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06254590
CAS Number: 610278-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR