3-Bromo-4-fluorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Bromo-4-fluorophenyl)boronic acid, 3-Bromo-4-fluorophenylboronic acid

19118-1G 19118-25G 19118-500MG 19118-5G
76909-570EA 153.84 USD
76909-570 76909-572 76909-574 76909-576
3-Bromo-4-fluorobenzeneboronic acid ≥97%
3-Bromo-4-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
MW: 218.816 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195642
CAS Number: 1092533-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR