3,5-Dibromo-2-fluoro-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3,5-dibromo-4-methylpyridine

19068-25G 19068-5G
76909-558EA 360.71 USD
76909-558 76909-560
3,5-Dibromo-2-fluoro-4-methylpyridine ≥97%
3,5-Dibromo-2-fluoro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₄NBr₂F
MW: 268.9100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864703
CAS Number: 1000340-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR