2-(Benzyloxy)-4-bromopyridine ≥95% research grade

Supplier: AOBChem USA

Warning

19026-1G 19026-25G 19026-5G
76909-548EA 94.43 USD
76909-548 76909-550 76909-552
2-(Benzyloxy)-4-bromopyridine ≥95% research grade
2-(Benzyloxy)-4-bromopyridine
Formula: C₁₂H₁₀BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD13190654
CAS Number: 960298-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR