3,4-Difluoro-2-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18923-25G 18923-5G
76909-526EA 519.85 USD
76909-526 76909-528
3,4-Difluoro-2-formylphenylboronic acid ≥97%
3,4-Difluoro-2-formylphenylboronic acid
Formula: C₇H₅BF₂O₃
MW: 185.921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713092
CAS Number: 1451392-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR