3,5-Dibromopyridin-4-ol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3,5-Dibromo-4-hydroxypyridine

18888-25G 18888-5G
76909-512EA 198.39 USD
76909-512 76909-514
3,5-Dibromopyridin-4-ol ≥97% research grade
3,5-Dibromopyridin-4-ol
Formula: C5H3Br2NO
Storage Temperature: Ambient
MDL Number: MFCD00082969
CAS Number: 25813-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR