3-Fluoro-2,4-dimethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18873-1G 18873-25G 18873-500MG 18873-5G
76909-496EA 379.81 USD
76909-496 76909-498 76909-500 76909-502
3-Fluoro-2,4-dimethylphenylboronic acid ≥97%
3-Fluoro-2,4-dimethylphenylboronic acid
Formula: C₈H₁₀BFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD23712905
CAS Number: 762286-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR