4-Bromo-2,6-diethylaniline ≥97%

Supplier: AOBChem USA

Warning

18689-1G 18689-25G 18689-5G
76909-456EA 62.6 USD
76909-456 76909-458 76909-460
4-Bromo-2,6-diethylaniline ≥97%
4-Bromo-2,6-diethylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrN
Storage Temperature: Ambient
MDL Number: MFCD00059266
CAS Number: 56746-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR