(3-Bromo-4-methylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-4-methylphenylboronic acid

18612-1G 18612-25G 18612-5G
76909-436EA 88.06 USD
76909-436 76909-438 76909-440
(3-Bromo-4-methylphenyl)boronic acid ≥95%
(3-Bromo-4-methylphenyl)boronic acid
Formula: C₇H₈BBrO₂
MW: 214.85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295252
CAS Number: 1312765-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR