3-Bromo-5-fluoro-2-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18608-25G 18608-5G
76909-426EA 418 USD
76909-426 76909-428
3-Bromo-5-fluoro-2-hydroxybenzaldehyde ≥97%
3-Bromo-5-fluoro-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
MW: 219.008 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458073
CAS Number: 178546-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR