3-Bromo-2-fluoro-6-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18596-1G 18596-500MG 18596-5G
76909-398EA 587.74 USD
76909-398 76909-400 76909-402
3-Bromo-2-fluoro-6-hydroxybenzaldehyde ≥97%
3-Bromo-2-fluoro-6-hydroxybenzaldehyde
Formula: C₇H₄BrFO₂
MW: 219.008 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15528068
CAS Number: 1160653-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR