3-Bromo-6-chloro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18590-1G 18590-25G 18590-5G
76909-370EA 108.22 USD
76909-370 76909-372 76909-374
3-Bromo-6-chloro-2-methoxybenzaldehyde ≥97%
3-Bromo-6-chloro-2-methoxybenzaldehyde
Formula: C₈H₆BrClO₂
MW: 249.489 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20528356
CAS Number: 1009094-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR