3,5-Dichloro-2-iodophenol ≥97%

Supplier: AOBChem USA

Warning

18483-1G 18483-25G 18483-500MG 18483-5G
76909-322EA 290.69 USD
76909-322 76909-324 76909-326 76909-328
3,5-Dichloro-2-iodophenol ≥97%
3,5-Dichloro-2-iodophenol
Formula: C₆H₃Cl₂IO
MW: 288.898 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12024395
CAS Number: 1028332-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR