3-Bromo-4-methyl-5-nitropyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-4-methy-5nitro-pyridine

18412-25G 18412-5G
76909-284EA 289.63 USD
76909-284 76909-286
3-Bromo-4-methyl-5-nitropyridine ≥97%
3-Bromo-4-methyl-5-nitropyridine
Formula: C6H5BrN2O2
MW: 217.0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847767
CAS Number: 69872-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR