4-(2-Bromobenzyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

18394-25G 18394-5G
76909-274EA 198.39 USD
76909-274 76909-276
4-(2-Bromobenzyl)morpholine ≥97%
4-(2-Bromobenzyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO
Storage Temperature: Ambient
MDL Number: MFCD00979294
CAS Number: 91130-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR