3,4-Dichloro-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18382-1G 18382-500MG 18382-5G
76909-262EA 765.97 USD
76909-262 76909-264 76909-266
3,4-Dichloro-2-fluorophenylboronic acid ≥97%
3,4-Dichloro-2-fluorophenylboronic acid
Formula: C₆H₄BCl₂FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD21602045
CAS Number: 1160561-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR