3-Bromo-5-fluoroisonicotinaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-fluoropyridine-4-carboxaldehyde

18215-1G 18215-25G 18215-5G
76909-216EA 81.69 USD
76909-216 76909-218 76909-220
3-Bromo-5-fluoroisonicotinaldehyde ≥97% research grade
3-Bromo-5-fluoroisonicotinaldehyde
Formula: C₆H₃BrFNO
MW: 204 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16606512
CAS Number: 1227573-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR