3-Cyano-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18113-1G 18113-25G 18113-500MG 18113-5G
76909-184EA 142.17 USD
76909-184 76909-186 76909-188 76909-190
3-Cyano-4-methoxyphenylboronic acid ≥97%
3-Cyano-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₃
MW: 176.965 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10697415
CAS Number: 911210-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR