3-Bromo-5-phenylpyridine ≥95%

Supplier: AOBChem USA

Warning

18098-1G 18098-25G 18098-5G
76909-172EA 84.88 USD
76909-172 76909-174 76909-176
3-Bromo-5-phenylpyridine ≥95%
3-Bromo-5-phenylpyridine
Formula: C₁₁H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD04114223
CAS Number: 142137-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR