3-Chloro-5-fluoro-4-methoxyphenol ≥95%

Supplier: AOBChem USA

Warning

18084-1G 18084-25G 18084-500MG 18084-5G
76909-158EA 147.47 USD
76909-158 76909-160 76909-162 76909-164
3-Chloro-5-fluoro-4-methoxyphenol ≥95%
3-Chloro-5-fluoro-4-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆ClFO₂
MW: 176.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832264
CAS Number: 1017777-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR