(3-Bromo-5-formylphenyl)boronic acid ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Bromo-5-formylphenylboronic acid
17798-1G
17798-25G
17798-5G
76909-126EA
94.43
USD
76909-126
76909-128
76909-130
(3-Bromo-5-formylphenyl)boronic acid ≥97% research grade
(3-Bromo-5-formylphenyl)boronic acid
Formula:
C₇H₆BBrO₃ MW: 228.83 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10696657 CAS Number: 157866-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...