5-Bromo-6-methoxynicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

17760-1G 17760-25G 17760-500MG 17760-5G
76909-100EA 159.14 USD
76909-100 76909-102 76909-104 76909-106
5-Bromo-6-methoxynicotinaldehyde ≥97%
5-Bromo-6-methoxynicotinaldehyde
Formula: C₇H₆BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16606522
CAS Number: 65873-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR