3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Formyl-5-(trifluoromethoxy)phenylboronic acid pinacol ester

17698-1G 17698-25G 17698-500MG 17698-5G
76909-064EA 160.2 USD
76909-064 76909-066 76909-068 76909-070
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethoxy)benzaldehyde ≥97%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethoxy)benzaldehyde
Formula: C₁₄H₁₆BF₃O₄
MW: 316.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181638
CAS Number: 1112209-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR