4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Dimethylcarbamoyl-2-fluorobenzeneboronic acid

17686-1G 17686-25G 17686-500MG 17686-5G
76909-050EA 194.15 USD
76909-050 76909-052 76909-054 76909-056
4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid ≥97%
4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BFNO₃
MW: 211.00 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11855833
CAS Number: 874289-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR