3-Bromo-5-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17660-25G 17660-5G
76909-030EA 524.09 USD
76909-030 76909-032
3-Bromo-5-(trifluoromethoxy)phenylboronic acid ≥97%
3-Bromo-5-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BBrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD11504853
CAS Number: 1072951-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR