3,5-Dibromoethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

17644-25G 17644-5G
76909-020EA 418 USD
76909-020 76909-022
3,5-Dibromoethoxybenzene ≥97%
3,5-Dibromoethoxybenzene
Formula: C₈H₈Br₂O
Storage Temperature: Ambient
MDL Number: MFCD17011980
CAS Number: 136265-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR