3-Bromo-2-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

17547-25G 17547-5G
76908-994EA 342.68 USD
76908-994 76908-996
3-Bromo-2-methylbenzaldehyde ≥95%
3-Bromo-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO
Storage Temperature: Refrigerator
MDL Number: MFCD11847533
CAS Number: 83647-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR