3-Chloro-5-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-5-(trifluoromethyl)benzeneboronic acid pinacol ester, 2-(3-Chloro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

17514-25G 17514-5G
76908-982EA 445.58 USD
76908-982 76908-984
3-Chloro-5-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%
3-Chloro-5-(trifluoromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11856039
CAS Number: 942069-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR