4-(1,3-Dioxolan-2-yl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

17457-25G 17457-5G
76908-974EA 453.01 USD
76908-974 76908-976
4-(1,3-Dioxolan-2-yl)benzonitrile ≥97%
4-(1,3-Dioxolan-2-yl)benzonitrile
Formula: C₁₀H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00052570
CAS Number: 66739-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR