3-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (3-Fluoro-5-(piperidine-1-carbonyl)phenyl)boronic acid

17334-1G 17334-25G 17334-500MG 17334-5G
76908-930EA 159.14 USD
76908-930 76908-932 76908-934 76908-936
3-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid ≥97% research grade
3-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid
Formula: C₁₂H₁₅BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08235060
CAS Number: 874219-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR