3-Fluor-5-(propylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-5-(propylcarbamoyl)phenylboronic acid

17330-25G 17330-5G
76908-920EA 595.17 USD
76908-920 76908-922
3-Fluor-5-(propylcarbamoyl)phenylboronic acid ≥97%
3-Fluor-5-(propylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD08235054
CAS Number: 874219-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR