4-(Methylthio)-2-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17320-1G 17320-25G 17320-5G
76908-884EA 62.6 USD
76908-884 76908-886 76908-888
4-(Methylthio)-2-(trifluoromethyl)phenylboronic acid ≥97%
4-(Methylthio)-2-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD09972107
CAS Number: 1072945-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR