3-Bromo-(5-methoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-(methoxycarbonyl)phenylboronic acid, (3-Bromo-5-(methoxycarbonyl)phenyl)boronic acid

17313-1G 17313-25G 17313-5G
76908-870EA 82.76 USD
76908-870 76908-872 76908-874
3-Bromo-(5-methoxycarbonyl)phenylboronic acid ≥97%
3-Bromo-(5-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BBrO₄
MW: 258.8617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689544
CAS Number: 913835-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR