3-Acetyl-2,6-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17210-1G 17210-25G 17210-500MG 17210-5G
76908-812EA 168.69 USD
76908-812 76908-814 76908-816 76908-818
3-Acetyl-2,6-difluorophenylboronic acid ≥97%
3-Acetyl-2,6-difluorophenylboronic acid
Formula: C₈H₇BF₂O₃
MW: 199.947 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11856033
CAS Number: 1451390-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR