3-Chloro-2,4-dimethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17174-1G 17174-500MG 17174-5G
76908-802EA 471.04 USD
76908-802 76908-804 76908-806
3-Chloro-2,4-dimethoxyphenylboronic acid ≥97%
3-Chloro-2,4-dimethoxyphenylboronic acid
Formula: C₈H₁₀BClO₄
MW: 216.427 g/mol
Storage Temperature: Ambient
CAS Number: 750585-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR