3-Ethoxy-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

17111-1G 17111-25G 17111-5G
76908-764EA 102.91 USD
76908-764 76908-766 76908-768
3-Ethoxy-2-fluorobenzoic acid ≥97%
3-Ethoxy-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD10699646
CAS Number: 1033201-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR