3-Bromo-2-chloro-5-fluorobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

17019-1G 17019-25G 17019-5G
76908-726EA 77.45 USD
76908-726 76908-728 76908-730
3-Bromo-2-chloro-5-fluorobenzotrifluoride ≥97%
3-Bromo-2-chloro-5-fluorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂BrClF₄
Storage Temperature: Ambient
MDL Number: MFCD09800681
CAS Number: 1027511-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR