3-Chloro-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

16850-25G 16850-5G
76908-664EA 183.54 USD
76908-664 76908-666
3-Chloro-2-methoxypyridine ≥97%
3-Chloro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD03095227
CAS Number: 13472-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR