3-Fluoro-5-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-5-hydroxybenzotrifluoride

16831-25G 16831-5G
76908-652EA 215.37 USD
76908-652 76908-654
3-Fluoro-5-(trifluoromethyl)phenol ≥97%
3-Fluoro-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄F₄O
MW: 180.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061288
CAS Number: 172333-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR