3-Bromo-2-hydroxy-5-methylbenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
16805-1G
16805-25G
16805-5G
76908-638EA
66.84
USD
76908-638
76908-640
76908-642
3-Bromo-2-hydroxy-5-methylbenzaldehyde ≥97%
3-Bromo-2-hydroxy-5-methylbenzaldehyde
Formula:
C₈H₇BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00275688 CAS Number: 33172-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...