(3-Chloro-6-fluoro-2-methoxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-6-fluoro-2-methoxyphenylboronic acid

16797-1G 16797-25G 16797-5G
76908-626EA 96.55 USD
76908-626 76908-628 76908-630
(3-Chloro-6-fluoro-2-methoxyphenyl)boronic acid ≥95%
(3-Chloro-6-fluoro-2-methoxyphenyl)boronic acid
Formula: C₇H₇BClFO₃
MW: 204.3911 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22414969
CAS Number: 2121511-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR