3-Fluoro-2-isopropoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16736-1G 16736-25G 16736-500MG 16736-5G
76908-608EA 225.98 USD
76908-608 76908-610 76908-612 76908-614
3-Fluoro-2-isopropoxyphenylboronic acid ≥97%
3-Fluoro-2-isopropoxyphenylboronic acid
Formula: C₉H₁₂BFO₃
MW: 197.999 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28360679
CAS Number: 2121511-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR