3-Chloro-2-formyl-4-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16716-1G 16716-25G 16716-500MG 16716-5G
76908-596EA 211.12 USD
76908-596 76908-598 76908-600 76908-602
3-Chloro-2-formyl-4-methylphenylboronic acid ≥97%
3-Chloro-2-formyl-4-methylphenylboronic acid
Formula: C₈H₈BClO₃
MW: 198.411 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181610
CAS Number: 1451391-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR