3-(Trifluoromethoxy)anisole ≥95%

Supplier: AOBChem USA

Warning

16668-25G 16668-5G
76908-578EA 346.92 USD
76908-578 76908-580
3-(Trifluoromethoxy)anisole ≥95%
3-(Trifluoromethoxy)anisole

Precaución: for Research Use Only

Formula: C₈H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD01631504
CAS Number: 142738-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR